(6-(Trifluoromethyl)pyridin-2-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A979423-1G A979423-250MG
77886-949EA 431 USD
77886-949 77886-950
(6-(Trifluoromethyl)pyridin-2-yl)boronic acid ≥95%
(6-(Trifluoromethyl)pyridin-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BF₃NO₂
MDL Number: MFCD08061586
CAS Number: 1162257-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR