(3S,6S,7R,7aR,8S,9S,10S,11aR,12R,12aR,13R,14R)-1-Ethyl-10,13-dimethoxy-3-(methoxymethyl)tetradecahydro-1H-3,6a,12-(epiethane[1,1,2]triyl)-7,9-methanonaphtho[2,3-b]azocine-6,8,11a-triol ≥98%

Supplier: Ambeed
Warning

Synonyms: Neoline

A978842-10MG A978842-1MG A978842-5MG
77687-002EA 65 USD
77687-002 77687-003 77687-004
(3S,6S,7R,7aR,8S,9S,10S,11aR,12R,12aR,13R,14R)-1-Ethyl-10,13-dimethoxy-3-(methoxymethyl)tetradecahydro-1H-3,6a,12-(epiethane[1,1,2]triyl)-7,9-methanonaphtho[2,3-b]azocine-6,8,11a-triol ≥98%
(3S,6S,7R,7aR,8S,9S,10S,11aR,12R,12aR,13R,14R)-1-Ethyl-10,13-dimethoxy-3-(methoxymethyl)tetradecahydro-1H-3,6a,12-(epiethane[1,1,2]triyl)-7,9-methanonaphtho[2,3-b]azocine-6,8,11a-triol

Precaución: for Research Use Only

Formula: C₂₄H₃₉NO₆
MW: 437.57 g/mol
MDL Number: MFCD01673484
CAS Number: 466-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR