3,4-Dihydro-2H-pyran-2-methanol ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxymethyl-3,4-dihydro-2H-pyran, DHP Linker

A978688-1G A978688-5G A978688-100G A978688-25G
77603-378EA 11.28 USD
77603-378 77603-380 77674-432 77674-434
3,4-Dihydro-2H-pyran-2-methanol ≥98%
3,4-Dihydro-2H-pyran-2-methanol

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD00051507
CAS Number: 3749-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR