1-Methyl-1H-indole-4-sulfonyl chloride ≥98%

Supplier: Ambeed
Danger

A977750-100MG A977750-250MG
77687-000EA 124 USD
77687-000 77687-001
1-Methyl-1H-indole-4-sulfonyl chloride ≥98%
1-Methyl-1H-indole-4-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₉H₈ClNO₂S
MDL Number: MFCD08690260
CAS Number: 876316-36-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR