[(R,R)-Teth-TsDPEN-RuCl] ≥98%

Supplier: Ambeed
Warning

Synonyms: Chloro[N-[(1R,2R)-1,2-diphenyl-2-[[3-(η6-phenyl)propyl]amino-κN]ethyl]-4-methylbenzenesulphonamidato-κN]ruthenium

A974987-1G A974987-250MG A974987-5G
77618-680EA 249.26 USD
77618-680 77747-731 77747-732
[(R,R)-Teth-TsDPEN-RuCl] ≥98%
[(R,R)-Teth-TsDPEN-RuCl]

Precaución: for Research Use Only

Formula: C₃₀H₃₁ClN₂O₂RuS
MW: 620.17 g/mol
MDL Number: MFCD16294993
CAS Number: 1192620-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR