Zinc(II) benzenesulfinate dihydrate ≥98%

Supplier: Ambeed
Warning

A974090-25G A974090-5G A974090-100G
77670-418EA 73 USD
77670-418 77670-420 77873-395
Zinc(II) benzenesulfinate dihydrate ≥98%
Zinc(II) benzenesulfinate dihydrate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₆S₂Zn
MDL Number: MFCD09702505
CAS Number: 47163-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR