N-((1R,2S,5R)-2-((S)-3-((7-(tert-Butyl)pyrazolo[1,5-a][1,3,5]triazin-4-yl)amino)-2-oxopyrrolidin-1-yl)-5-(tert-butylamino)cyclohexyl)acetamide ≥97%
Supplier: Ambeed
|
Warning
|
A973172-1MG
A973172-5MG
A973172-10MG
77599-326EA
26.65
USD
77599-326
77599-328
77623-226
N-((1R,2S,5R)-2-((S)-3-((7-(tert-Butyl)pyrazolo[1,5-a][1,3,5]triazin-4-yl)amino)-2-oxopyrrolidin-1-yl)-5-(tert-butylamino)cyclohexyl)acetamide ≥97%
N-((1R,2S,5R)-2-((S)-3-((7-(tert-Butyl)pyrazolo[1,5-a][1,3,5]triazin-4-yl)amino)-2-oxopyrrolidin-1-yl)-5-(tert-butylamino)cyclohexyl)acetamide
Precaución: for Research Use Only
Formula:
C₂₅H₄₀N₈O₂ |
MDL Number:
MFCD31540731 CAS Number: 1286279-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...