1,3-Bis(dicyanomethylidene)indan ≥98%

Supplier: Ambeed
Danger

A971066-1G A971066-250MG
77784-452EA 57 USD
77784-452 77784-453
1,3-Bis(dicyanomethylidene)indan ≥98%
1,3-Bis(dicyanomethylidene)indan

Precaución: for Research Use Only

Formula: C₁₅H₆N₄
MDL Number: MFCD00142514
CAS Number: 38172-19-9
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR