3-Chloro-5-(trifluoromethyl)pyridazine ≥98%

Supplier: Ambeed
Warning

A970845-1G A970845-250MG
77886-937EA 405 USD
77886-937 77886-938
3-Chloro-5-(trifluoromethyl)pyridazine ≥98%
3-Chloro-5-(trifluoromethyl)pyridazine

Precaución: for Research Use Only

Formula: C₅H₂ClF₃N₂
MDL Number: MFCD20275278
CAS Number: 845617-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR