NVP-BEP800 ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-(2,4-dichloro-5-(2-(pyrrolidin-1-yl)ethoxy)phenyl)-N-ethylthieno[2,3-d]pyrimidine-6-carboxamide, VER-82576, 2-amino-4-[2,4-dichloro-5-[2-(1-pyrrolidinyl)ethoxy]phenyl]-N-ethyl-thieno[2,3-d]pyrimidine-6-carboxamide

A969983-10MG A969983-1MG A969983-5MG
77618-664EA 94.76 USD
77618-664 77618-666 77618-668
NVP-BEP800 ≥98%
NVP-BEP800

Precaución: for Research Use Only

Formula: C₂₁H₂₃Cl₂N₅O₂S
MDL Number: MFCD18251568
CAS Number: 847559-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR