tert-Butyl 4-oxobutanoate ≥97%

Supplier: Ambeed
Warning

A969806-1G A969806-250MG A969806-5G
77684-904EA 150 USD
77684-904 77684-905 77684-906
tert-Butyl 4-oxobutanoate ≥97%
tert-Butyl 4-oxobutanoate

Precaución: For research use only.

Formula: C₈H₁₄O₃
MW: 158.2 g/mol
MDL Number: MFCD11975807
CAS Number: 51534-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR