6-(Difluoro(6-(pyridin-4-yl)-[1,2,4]triazolo[4,3-b]pyridazin-3-yl)methyl)quinoline ≥98%

Supplier: Ambeed
Warning

Synonyms: JNJ-38877618

A967658-10MG A967658-1MG A967658-5MG
77599-318EA 208.08 USD
77599-318 77599-320 77599-322
6-(Difluoro(6-(pyridin-4-yl)-[1,2,4]triazolo[4,3-b]pyridazin-3-yl)methyl)quinoline ≥98%
6-(Difluoro(6-(pyridin-4-yl)-[1,2,4]triazolo[4,3-b]pyridazin-3-yl)methyl)quinoline

Precaución: for Research Use Only

Formula: C₂₀H₁₂F₂N₆
MW: 374.35 g/mol
MDL Number: MFCD31657377
CAS Number: 943540-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR