tert-Butyl (5-(methylamino)pentyl)carbamate ≥95%

Supplier: Ambeed
Danger

Synonyms: 5-(Methylamino)-N-Boc-pentanamine

A967545-100MG A967545-250MG A967545-50MG A967545-5G A967545-1G
77599-314EA 36.9 USD
77599-314 77599-316 77623-218 77784-446 77784-445
tert-Butyl (5-(methylamino)pentyl)carbamate ≥95%
tert-Butyl (5-(methylamino)pentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₄N₂O₂
MW: 216.3205 g/mol
MDL Number: MFCD28148216
CAS Number: 1311458-36-2
UN: 3082
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR