1H-Indole-2-carboxamide ≥95%

Supplier: Ambeed
Warning

A967498-250MG A967498-100MG A967498-1G
77603-368EA 34.85 USD
77603-368 77670-406 77880-784
1H-Indole-2-carboxamide ≥95%
1H-Indole-2-carboxamide

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MDL Number: MFCD00231036
CAS Number: 1670-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR