6-Bromonaphthalen-1-amine ≥98%

Supplier: Ambeed
Warning

A967179-100MG A967179-250MG
77614-686EA 56.65 USD
77614-686 77884-902
6-Bromonaphthalen-1-amine ≥98%
6-Bromonaphthalen-1-amine

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MDL Number: MFCD17012559
CAS Number: 591253-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR