3-Bromo-5-chloro-4-methoxypyridine ≥95%

Supplier: Ambeed
Warning

A966493-100MG A966493-1G A966493-250MG
77684-901EA 164 USD
77684-901 77684-902 77684-903
3-Bromo-5-chloro-4-methoxypyridine ≥95%
3-Bromo-5-chloro-4-methoxypyridine

Precaución: For research use only.

Formula: C₆H₅BrClNO
MW: 222.47 g/mol
MDL Number: MFCD13185355
CAS Number: 1214362-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR