Fmoc-3-methoxy-L-homophenylalanine ≥95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-HoPhe(3-OMe)-OH

A966021-250MG A966021-50MG A966021-1G A966021-100MG
77618-652EA 111.65 USD
77618-652 77618-654 77749-799 77749-798
Fmoc-3-methoxy-L-homophenylalanine ≥95%
Fmoc-3-methoxy-L-homophenylalanine

Precaución: for Research Use Only

Formula: C₂₆H₂₅NO₅
MDL Number: MFCD07372234
CAS Number: 1260610-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR