2-Hydroxy-4-methoxy-4'-methylbenzophenone ≥98%

Supplier: Ambeed
Warning

Synonyms: (2-Hydroxy-4-methoxyphenyl)(p-tolyl)methanone, Mexenone

A966002-100MG A966002-1G A966002-250MG
77618-646EA 49.44 USD
77618-646 77618-648 77618-650
2-Hydroxy-4-methoxy-4'-methylbenzophenone ≥98%
2-Hydroxy-4-methoxy-4'-methylbenzophenone

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.2699 g/mol
MDL Number: MFCD00017172
CAS Number: 1641-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR