2-Methoxy-1-propyl acetate ≥95%

Supplier: Ambeed
Danger

A965114-1G A965114-25G
77623-212EA 18.54 USD
77623-212 77623-214
2-Methoxy-1-propyl acetate ≥95%
2-Methoxy-1-propyl acetate

Precaución: for Research Use Only

Formula: C₆H₁₂O₃
MDL Number: MFCD00071665
CAS Number: 70657-70-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR