2-[(Trifluoromethyl)sulfanyl]-1,2-benzothiazol-3(2H)-one 1,1-dioxide ≥98%

Supplier: Ambeed
Danger

A965098-5G A965098-100MG A965098-1G
77599-310EA 416.15 USD
77599-310 77784-438 77784-439
2-[(Trifluoromethyl)sulfanyl]-1,2-benzothiazol-3(2H)-one 1,1-dioxide ≥98%
2-[(Trifluoromethyl)sulfanyl]-1,2-benzothiazol-3(2H)-one 1,1-dioxide

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₃S₂
MDL Number: MFCD28127153
CAS Number: 1647073-46-8
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR