(5S,8S,11S,12R)-11-((S)-sec-Butyl)-1-(9H-fluoren-9-yl)-5,8-diisopropyl-12-methoxy-4,10-dimethyl-3,6,9-trioxo-2-oxa-4,7,10-triazatetradecan-14-oic acid ≥98%

Supplier: Ambeed
Warning

A964901-100MG A964901-50MG A964901-250MG
77623-206EA 77.45 USD
77623-206 77623-208 77784-435
(5S,8S,11S,12R)-11-((S)-sec-Butyl)-1-(9H-fluoren-9-yl)-5,8-diisopropyl-12-methoxy-4,10-dimethyl-3,6,9-trioxo-2-oxa-4,7,10-triazatetradecan-14-oic acid ≥98%
(5S,8S,11S,12R)-11-((S)-sec-Butyl)-1-(9H-fluoren-9-yl)-5,8-diisopropyl-12-methoxy-4,10-dimethyl-3,6,9-trioxo-2-oxa-4,7,10-triazatetradecan-14-oic acid

Precaución: for Research Use Only

Formula: C₃₆H₅₁N₃O₇
MDL Number: MFCD28386982
CAS Number: 863971-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR