3-Butylisobenzofuran-1(3H)-one ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-Butylphthalide

A963975-250MG A963975-1G A963975-100MG
77749-796EA 193 USD
77749-796 77749-795 77749-794
3-Butylisobenzofuran-1(3H)-one ≥98%
3-Butylisobenzofuran-1(3H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₂
MDL Number: MFCD01704513
CAS Number: 6066-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR