2,5-Dioxopyrrolidin-1-yl 1-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13,16,19-pentaoxa-4-azadocosan-22-oate ≥98%

Supplier: Ambeed
Warning

Synonyms: Mal-amido-PEG5-C2-NHS ester

A961305-100MG A961305-250MG A961305-1G
77614-676EA 73.13 USD
77614-676 77614-678 77686-989
2,5-Dioxopyrrolidin-1-yl 1-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13,16,19-pentaoxa-4-azadocosan-22-oate ≥98%
2,5-Dioxopyrrolidin-1-yl 1-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13,16,19-pentaoxa-4-azadocosan-22-oate

Precaución: for Research Use Only

Formula: C₂₄H₃₅N₃O₁₂
MW: 557.55 g/mol
MDL Number: MFCD20230002
CAS Number: 1315355-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR