2-(Benzyl(phenyl)amino)ethyl 5-(5,5-dimethyl-2-oxido-1,3,2-dioxaphosphinan-2-yl)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3-carboxylate ≥98%

Supplier: Ambeed
Danger

Synonyms: Efonidipine

A960939-5MG A960939-100MG A960939-10MG A960939-2MG
77614-674EA 28.84 USD
77614-674 77686-986 77686-987 77686-988
2-(Benzyl(phenyl)amino)ethyl 5-(5,5-dimethyl-2-oxido-1,3,2-dioxaphosphinan-2-yl)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3-carboxylate ≥98%
2-(Benzyl(phenyl)amino)ethyl 5-(5,5-dimethyl-2-oxido-1,3,2-dioxaphosphinan-2-yl)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₃₄H₃₈N₃O₇P
MDL Number: MFCD00865899
CAS Number: 111011-63-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR