Pyridin-3-yl isocyanate ≥97%

Supplier: Ambeed
Danger

A960876-100MG A960876-250MG
77882-847EA 95 USD
77882-847 77882-848
Pyridin-3-yl isocyanate ≥97%
Pyridin-3-yl isocyanate

Precaución: for Research Use Only

Formula: C₆H₄N₂O
MDL Number: MFCD01075747
CAS Number: 15268-31-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR