6-Azaspiro[3.4]octane ≥98%

Supplier: Ambeed
Danger

A960125-100MG A960125-1G A960125-250MG
77891-721EA 98 USD
77891-721 77891-722 77891-723
6-Azaspiro[3.4]octane ≥98%
6-Azaspiro[3.4]octane

Precaución: for Research Use Only

Formula: C₇H₁₃N
MDL Number: MFCD09759054
CAS Number: 765-64-0
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR