1-Pyrenebutanoic acid Succinimidyl ester ≥98%

Supplier: Ambeed
Warning

A959894-1G A959894-250MG A959894-100MG A959894-5G
77603-356EA 73.8 USD
77603-356 77614-670 77686-985 77902-059
1-Pyrenebutanoic acid Succinimidyl ester ≥98%
1-Pyrenebutanoic acid Succinimidyl ester

Precaución: for Research Use Only

Formula: C₂₄H₁₉NO₄
MDL Number: MFCD00077403
CAS Number: 114932-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR