Biotin-PEG4-NHS ≥97%

Supplier: Ambeed
Warning

A959877-50MG A959877-100MG A959877-250MG
77599-306EA 39.98 USD
77599-306 77623-188 77623-190
Biotin-PEG4-NHS ≥97%
Biotin-PEG4-NHS

Precaución: for Research Use Only

Formula: C₂₅H₄₀N₄O₁₀S
MDL Number: MFCD08457831
CAS Number: 459426-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR