4'-Aminopropiophenone ≥98%

Supplier: Ambeed
Danger

Synonyms: 1-(4-Aminophenyl)propan-1-one, p-Aminopropiophenone

A959052-1G A959052-5G
77747-715EA 45 USD
77747-715 77747-716
4'-Aminopropiophenone ≥98%
4'-Aminopropiophenone

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.19 g/mol
MDL Number: MFCD00017113
CAS Number: 70-69-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR