cis-Cyclobutane-1,3-diol ≥97%

Supplier: Ambeed
Warning

A958050-250MG A958050-1G
77749-789EA 242 USD
77749-789 77749-788
cis-Cyclobutane-1,3-diol ≥97%
cis-Cyclobutane-1,3-diol

Precaución: for Research Use Only

Formula: C₄H₈O₂
MDL Number: MFCD28506109
CAS Number: 1332482-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR