rel-2-((1r,4r)-4-(4-(2-((4-(Trifluoromethoxy)benzyl)carbamoyl)imidazo[1,2-a]pyridin-6-yl)phenyl)cyclohexyl)acetic acid ≥98%

Supplier: Ambeed
Warning

A957858-100MG A957858-50MG A957858-1G A957858-250MG
77614-666EA 322.39 USD
77614-666 77614-668 77686-983 77686-984
rel-2-((1r,4r)-4-(4-(2-((4-(Trifluoromethoxy)benzyl)carbamoyl)imidazo[1,2-a]pyridin-6-yl)phenyl)cyclohexyl)acetic acid ≥98%
rel-2-((1r,4r)-4-(4-(2-((4-(Trifluoromethoxy)benzyl)carbamoyl)imidazo[1,2-a]pyridin-6-yl)phenyl)cyclohexyl)acetic acid

Precaución: for Research Use Only

Formula: C₃₀H₂₈F₃N₃O₄
MDL Number: MFCD28167832
CAS Number: 1449779-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR