4-(Ethoxycarbonylmethyl)phenylboronic acid ≥96%

Supplier: Ambeed
Warning

Synonyms: 4-[(Ethoxycarbonyl)methyl]phenylboronic acid, (4-(2-Ethoxy-2-oxoethyl)phenyl)boronic acid

A957827-100MG A957827-1G A957827-250MG A957827-5G
77873-387EA 81 USD
77873-387 77873-388 77873-389 77873-390
4-(Ethoxycarbonylmethyl)phenylboronic acid ≥96%
4-(Ethoxycarbonylmethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₄
MDL Number: MFCD08704683
CAS Number: 92243-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR