3-(5-Bromo-furan-2-yl)-acrylic acid ≥95%

Supplier: Ambeed
Warning

A957733-5G A957733-100MG A957733-250MG
77686-982EA 110 USD
77686-982 77902-053 77901-009
3-(5-Bromo-furan-2-yl)-acrylic acid ≥95%
3-(5-Bromo-furan-2-yl)-acrylic acid

Precaución: for Research Use Only

Formula: C₇H₅BrO₃
MDL Number: MFCD02708232
CAS Number: 64154-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR