Phenyl ethenesulfonate ≥97% stabilised

Supplier: Ambeed
Warning


Stabilization: Stabilised with TBC

A957616-1G A957616-250MG A957616-100MG A957616-5G
77670-400EA 116 USD
77670-400 77670-402 77880-779 77880-780
Phenyl ethenesulfonate ≥97% stabilised
Phenyl ethenesulfonate

Precaución: for Research Use Only

Formula: C₈H₈O₃S
MW: 184.21 g/mol
MDL Number: MFCD00012206
CAS Number: 1562-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR