N-tert-Butylaniline ≥90%

Supplier: Ambeed
Warning

A957490-250MG A957490-100MG
77623-186EA 147.29 USD
77623-186 77784-416
N-tert-Butylaniline ≥90%
N-tert-Butylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₅N
MDL Number: MFCD05669577
CAS Number: 937-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR