4-Fluoropicolinic acid ≥97%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A955190-1G
A955190-5G
A955190-250MG
77573-892EA
229.69
USD
77573-892
77614-662
77684-898
4-Fluoropicolinic acid ≥97%, Ambeed.Inc
4-Fluoropicolinic acid
Precaución: For research use only.
Formula:
C₆H₄FNO₂ MW: 141.0999 g/mol |
MDL Number:
MFCD07374895 CAS Number: 886371-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...