N-(4-((3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)amino)quinazolin-6-yl)acrylamide 4-methylbenzenesulfonate ≥99%

Supplier: Ambeed
Warning

A954947-1MG A954947-5MG A954947-10MG A954947-50MG
77603-348EA 16.4 USD
77603-348 77603-350 77618-634 77749-785
N-(4-((3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)amino)quinazolin-6-yl)acrylamide 4-methylbenzenesulfonate ≥99%
N-(4-((3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)amino)quinazolin-6-yl)acrylamide 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₃₁H₂₆ClFN₄O₅S
MDL Number: MFCD17215204
CAS Number: 1050500-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR