Disodium clodronate tetrahydrate ≥99%

Supplier: Ambeed
Danger

A954132-5G A954132-1G A954132-25G
77627-780EA 54.59 USD
77627-780 77880-776 77880-777
Disodium clodronate tetrahydrate ≥99%
Disodium clodronate tetrahydrate

Precaución: for Research Use Only

Formula: CH₄Cl₂O₆P₂
MDL Number: MFCD02940846
CAS Number: 88416-50-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR