3-(1-Methyl-4-morpholino-1H-pyrazolo[3,4-d]pyrimidin-6-yl)phenol ≥99%

Supplier: Ambeed
Warning

Synonyms: ETP-45658

A952617-250MG A952617-100MG A952617-50MG
77749-781EA 466 USD
77749-781 77749-780 77749-782
3-(1-Methyl-4-morpholino-1H-pyrazolo[3,4-d]pyrimidin-6-yl)phenol ≥99%
3-(1-Methyl-4-morpholino-1H-pyrazolo[3,4-d]pyrimidin-6-yl)phenol

Precaución: for Research Use Only

Formula: C₁₆H₁₇N₅O₂
MW: 311.34 g/mol
MDL Number: MFCD29081201
CAS Number: 1198357-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR