2,5-Dichloro-4-nitropyridine ≥97%

Supplier: Ambeed
Warning

A952090-1G A952090-100MG
77618-632EA 951.72 USD
77618-632 77747-710
2,5-Dichloro-4-nitropyridine ≥97%
2,5-Dichloro-4-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂N₂O₂
MDL Number: MFCD28648163
CAS Number: 1400701-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR