8-Phenylcaprylic acid ≥98%

Supplier: Ambeed
Warning

A951916-100MG A951916-5G
77878-753EA 42 USD
77878-753 77878-754
8-Phenylcaprylic acid ≥98%
8-Phenylcaprylic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₂
MDL Number: MFCD00037802
CAS Number: 26547-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR