Tetrakis(4-cyanophenyl)methane ≥97%

Supplier: Ambeed
Warning

A950737-100MG A950737-250MG
77618-630EA 24.72 USD
77618-630 77749-779
Tetrakis(4-cyanophenyl)methane ≥97%
Tetrakis(4-cyanophenyl)methane

Precaución: for Research Use Only

Formula: C₂₉H₁₆N₄
MDL Number: MFCD29049230
CAS Number: 121706-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR