Takinib ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-N-(1-Propylbenzimidazol-2-yl)benzene-1,3-dicarboxamide, N1-(1-Propyl-1H-benzo[d]imidazol-2-yl)isophthalamide

A949172-10MG A949172-1MG A949172-5MG
77599-294EA 108.65 USD
77599-294 77599-296 77599-298
Takinib ≥98%
Takinib

Precaución: for Research Use Only

Formula: C₁₈H₁₈N₄O₂
MW: 322.36 g/mol
MDL Number: MFCD11822633
CAS Number: 1111556-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR