tert-Butyl (4-(2-hydroxypropan-2-yl)phenyl)carbamate ≥98%

Supplier: Ambeed
Warning

A948698-100MG A948698-1G
77784-398EA 30 USD
77784-398 77784-399
tert-Butyl (4-(2-hydroxypropan-2-yl)phenyl)carbamate ≥98%
tert-Butyl (4-(2-hydroxypropan-2-yl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO₃
MDL Number: MFCD30735824
CAS Number: 1787261-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR