3-Bromo-4-fluoro-1H-pyrazole ≥98%
Supplier: Ambeed
|
Warning
|
A948680-250MG
A948680-100MG
77784-397EA
158
USD
77784-397
77784-396
3-Bromo-4-fluoro-1H-pyrazole ≥98%
3-Bromo-4-fluoro-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₃H₂BrFN₂ |
MDL Number:
MFCD20926086 CAS Number: 1621526-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...