3-Amino-5-chloro-1-methyl-2(1H)-pyridinone ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Amino-5-chloro-1-methylpyridin-2(1H)-one
A946382-1G
A946382-250MG
A946382-5G
77684-893EA
66
USD
77684-893
77684-894
77684-895
3-Amino-5-chloro-1-methyl-2(1H)-pyridinone ≥98%
3-Amino-5-chloro-1-methyl-2(1H)-pyridinone
Precaución: For research use only.
Formula:
C₆H₇ClN₂O MW: 158.586 g/mol |
MDL Number:
MFCD28130270 CAS Number: 1441769-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...