(2R,3S)-1-Benzhydryl-2-methylazetidin-3-ol ≥98%

Supplier: Ambeed
Warning

A946357-5G A946357-250MG A946357-1G
77618-608EA 212.18 USD
77618-608 77749-776 77749-775
(2R,3S)-1-Benzhydryl-2-methylazetidin-3-ol ≥98%
(2R,3S)-1-Benzhydryl-2-methylazetidin-3-ol

Precaución: for Research Use Only

Formula: C₁₇H₁₉NO
MW: 253.34 g/mol
MDL Number: MFCD29059120
CAS Number: 138876-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR