Methyl 3-(Hydroxymethyl)-5-nitrobenzoate ≥95%

Supplier: Ambeed
Warning

A945919-100MG A945919-250MG
77880-766EA 9 USD
77880-766 77880-767
Methyl 3-(Hydroxymethyl)-5-nitrobenzoate ≥95%
Methyl 3-(Hydroxymethyl)-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₉NO₅
MDL Number: MFCD08275104
CAS Number: 53732-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR