(3R)-3-(Bromomethyl)pyrrolidine, N-BOC protected ≥96%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl (3R)-3-(bromomethyl)pyrrolidine-1-carboxylate, (R)-1-Boc-3-(Bromomethyl)pyrrolidine
A945400-250MG
A945400-100MG
A945400-5G
77614-660EA
92.7
USD
77614-660
77684-891
77684-892
(3R)-3-(Bromomethyl)pyrrolidine, N-BOC protected ≥96%
(3R)-3-(Bromomethyl)pyrrolidine, N-BOC protected
Precaución: For research use only.
Formula:
C₁₀H₁₈BrNO₂ MW: 264.159 g/mol |
MDL Number:
MFCD08059335 CAS Number: 1067230-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...