Methyl-6-(trifluoromethyl)-1H-indole-2-carboxylate ≥95%

Supplier: Ambeed
Warning

A945056-100MG A945056-250MG
77884-889EA 32 USD
77884-889 77884-890
Methyl-6-(trifluoromethyl)-1H-indole-2-carboxylate ≥95%
Methyl-6-(trifluoromethyl)-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₈F₃NO₂
MDL Number: MFCD07357592
CAS Number: 887360-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR